siRNA Details
| Gene Symbol: PAQR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017022364.1 | XP_016877853.1 | 1 | 117 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000558269.2 | - | 3 | 322 | Chr. 15 - 69278675 - 69288564 [+] on Build GRCh38 | |

PAQR5
Genomic Map