siRNA Details
Gene Symbol: LINC01583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120367.1 | 5 | 2164 | |||
GenBank mRNA | BC043257.2 | - | 5 | 2164 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000558082.1 | - | 5 | 2164 | Chr. 15 - 82088594 - 82097694 [+] on Build GRCh38 |
