siRNA Details
| Gene Symbol: LINC01583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120367.1 | 5 | 2098 | |||
| GenBank mRNA | BC043257.2 | - | 5 | 2098 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000558082.1 | - | 5 | 2098 | Chr. 15 - 82088594 - 82097694 [+] on Build GRCh38 | |

LINC01583
Genomic Map