siRNA Details
| Gene Symbol: LOC101928414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120335.1 | 4 | 134 | |||
| GenBank mRNA | HY107468.1 | - | 4 | 137 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928414| Gene Symbol: LOC101928414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120335.1 | 4 | 134 | |||
| GenBank mRNA | HY107468.1 | - | 4 | 137 | ||