siRNA Details
| Gene Symbol: LOC102723335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135693.1 | 2 | 227 | |||
| GenBank mRNA | BC032026.1 | - | 2 | 217 | ||
| CR748292.1 | - | 2 | 276 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723335| Gene Symbol: LOC102723335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135693.1 | 2 | 227 | |||
| GenBank mRNA | BC032026.1 | - | 2 | 217 | ||
| CR748292.1 | - | 2 | 276 | |||