siRNA Details
| Gene Symbol: LOC101927751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135828.1 | 2 | 1480 | |||
| NR_135831.1 | 2 | 2396 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000560068.1 | - | 2 | 1435 | Chr. 15 - 100913203 - 100919238 [-] on Build GRCh38 | |

LOC101927751
Genomic Map