siRNA Details
| Gene Symbol: LOC102724452 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135683.1 | 2 | 179 | |||
| GenBank mRNA | BX100197.1 | - | 2 | 179 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000558587.1 | - | 2 | 179 | Chr. 15 - 86938797 - 86988426 [-] on Build GRCh38 | |

LOC102724452
Genomic Map