siRNA Details
| Gene Symbol: LOC100128108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321551.1 | NP_001308480.1 | 3 | 343 | ||
| NM_001321552.1 | NP_001308481.1 | 3 | 357 | |||
| NM_001321553.1 | NP_001308482.1 | 2 | 426 | |||
| XM_006720812.3 | XP_006720875.1 | 3 | 1347 | |||
| XM_017021852.1 | XP_016877341.1 | 3 | 684 | |||
| GenBank mRNA | HY056262.1 | - | 3 | 357 | ||

LOC100128108
Genomic Map