siRNA Details
| Gene Symbol: LOC100128108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321553.1 | NP_001308482.1 | 1 | 14 | ||
| XM_006720812.3 | XP_006720875.1 | 1 | 873 | |||
| Gene Symbol: LOC400464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127420.1 | - | 2 | 922 | ||

LOC100128108
Genomic Map