siRNA Details
| Gene Symbol: LOC100131315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109767.1 | 1 | 194 | |||
| GenBank mRNA | AW582922.1 | - | 1 | 194 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000560363.1 | - | 1 | 183 | Chr. 15 - 32718101 - 32718854 [-] on Build GRCh38 | |

LOC100131315
Genomic Map