siRNA Details
Gene Symbol: LINC01581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120320.1 | 6 | 1986 | |||
GenBank mRNA | BC043440.2 | - | 6 | 1986 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000558874.1 | - | 6 | 1986 | Chr. 15 - 93905405 - 94107938 [-] on Build GRCh38 |
