siRNA Details
| Gene Symbol: LOC102724034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120378.1 | 3 | 483 | |||
| GenBank mRNA | DC422276.1 | - | 3 | 483 | ||
| Gene Symbol: LOC103171574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120379.1 | 3 | 582 | |||
| GenBank mRNA | AK311032.1 | - | 3 | 483 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000559866.1 | - | 3 | 483 | Chr. 15 - 84422618 - 84425882 [+] on Build GRCh38 | |

LOC102724034
Genomic Map