siRNA Details
| Gene Symbol: LOC102723640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135684.1 | 4 | 466 | |||
| NR_135685.1 | 3 | 348 | ||||
| GenBank mRNA | BG203382.1 | - | 1 | 113 | ||
| HY009132.1 | - | 4 | 467 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723640| Gene Symbol: LOC102723640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135684.1 | 4 | 466 | |||
| NR_135685.1 | 3 | 348 | ||||
| GenBank mRNA | BG203382.1 | - | 1 | 113 | ||
| HY009132.1 | - | 4 | 467 | |||