siRNA Details
| Gene Symbol: LINC01566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027079.2 | 9 | 2550 | |||
| GenBank mRNA | BC045579.1 | - | 9 | 2550 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000569242.1 | - | 9 | 2550 | Chr. 16 - 35363531 - 35390584 [+] on Build GRCh38 | |

LINC01566
Genomic Map