siRNA Details
| Gene Symbol: LOC101927793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135194.1 | 1 | 52 | |||
| GenBank mRNA | AK055866.1 | - | 1 | 295 | ||
| DA400871.1 | - | 2 | 408 | |||
| DA641322.1 | - | 2 | 179 | |||
| DB173038.1 | - | 2 | 221 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927793| Gene Symbol: LOC101927793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135194.1 | 1 | 52 | |||
| GenBank mRNA | AK055866.1 | - | 1 | 295 | ||
| DA400871.1 | - | 2 | 408 | |||
| DA641322.1 | - | 2 | 179 | |||
| DB173038.1 | - | 2 | 221 | |||