siRNA Details
| Gene Symbol: LOC158402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK095652.1 | - | 1 | 211 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000563434.1 | - | 1 | 299 | Chr. 9 - 112032555 - 112037730 [-] on Build GRCh38 | |

LOC158402
Genomic Map