siRNA Details
Gene Symbol: LOC105371925 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135654.1 | 1 | 109 | |||
GenBank mRNA | AL832593.1 | - | 1 | 109 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000575922.5 | - | 1 | 109 | Chr. 17 - 81228707 - 81233983 [+] on Build GRCh38 |
