siRNA Details
| Gene Symbol: LOC105447648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131191.1 | 2 | 466 | |||
| GenBank mRNA | BX458411.2 | - | 2 | 466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105447648| Gene Symbol: LOC105447648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131191.1 | 2 | 466 | |||
| GenBank mRNA | BX458411.2 | - | 2 | 466 | ||