siRNA Details
| Gene Symbol: LOC101928614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135181.1 | 2 | 390 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000563331.1 | - | 2 | 385 | Chr. 16 - 86720688 - 86721954 [-] on Build GRCh38 | |

LOC101928614
Genomic Map