siRNA Details
| Gene Symbol: LINC01070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CR627049.1 | - | 2 | 188 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000561786.1 | - | 2 | 187 | Chr. 13 - 112197350 - 112201000 [+] on Build GRCh38 | |

LINC01070
Genomic Map