siRNA Details
Gene Symbol: LOC101927272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110908.1 | 3 | 990 | |||
GenBank mRNA | AF143871.1 | - | 3 | 999 | ||
AL161979.1 | - | 2 | 5156 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110908.1 | 3 | 990 | |||
GenBank mRNA | AF143871.1 | - | 3 | 999 | ||
AL161979.1 | - | 2 | 5156 |