siRNA Details
| Gene Symbol: LINC00919 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038233.1 | 3 | 732 | |||
| GenBank mRNA | AK098676.1 | - | 3 | 732 | ||
| BC105278.1 | - | 2 | 620 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00919| Gene Symbol: LINC00919 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038233.1 | 3 | 732 | |||
| GenBank mRNA | AK098676.1 | - | 3 | 732 | ||
| BC105278.1 | - | 2 | 620 | |||