siRNA Details
| Gene Symbol: LOC105371414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136339.1 | 3 | 1484 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000563087.2 | - | 2 | 318 | Chr. 16 - 89297508 - 89298309 [+] on Build GRCh38 | |

LOC105371414
Genomic Map