siRNA Details
Gene Symbol: LINC01570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110902.1 | 3 | 652 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000566218.1 | - | 3 | 598 | Chr. 16 - 5601169 - 5616196 [-] on Build GRCh38 |
