siRNA Details
| Gene Symbol: LINC01570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110902.1 | 2 | 201 | |||
| GenBank mRNA | DB449025.1 | - | 2 | 121 | ||
| HY009375.1 | - | 2 | 196 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000566218.1 | - | 2 | 147 | Chr. 16 - 5601169 - 5616196 [-] on Build GRCh38 | |

LINC01570
Genomic Map