siRNA Details
| Gene Symbol: LOC101928880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110944.1 | 2, 3 | 350 | |||
| GenBank mRNA | BX110403.1 | - | 2, 3 | 350 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000569362.2 | - | 3 | 350 | Chr. 16 - 88194314 - 88195217 [+] on Build GRCh38 | |

LOC101928880
Genomic Map