siRNA Details
Gene Symbol: LOC101928880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110944.1 | 2 | 268 | |||
GenBank mRNA | BX110403.1 | - | 2 | 268 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000569362.2 | - | 2 | 268 | Chr. 16 - 88194314 - 88195217 [+] on Build GRCh38 |
