siRNA Details
Gene Symbol: LOC101930452 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120479.1 | 1 | 1117 | |||
GenBank mRNA | BC040314.1 | - | 1 | 1093 | ||
BC047234.1 | - | 1 | 1117 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000567749.1 | - | 1 | 1117 | Chr. 12 - 9367464 - 9397617 [+] on Build GRCh38 |
