siRNA Details
| Gene Symbol: LINC01544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103449.1 | 4 | 822 | |||
| GenBank mRNA | AK054836.1 | - | 4 | 822 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000567801.2 | - | 4 | 822 | Chr. 18 - 61748176 - 61754695 [+] on Build GRCh38 | |

LINC01544
Genomic Map