siRNA Details
| Gene Symbol: LCMT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120306.1 | 1 | 72 | |||
| GenBank mRNA | AL600125.1 | - | 1 | 73 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LCMT1-AS1| Gene Symbol: LCMT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120306.1 | 1 | 72 | |||
| GenBank mRNA | AL600125.1 | - | 1 | 73 | ||