siRNA Details
| Gene Symbol: LINC01290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BF853722.1 | - | 2 | 312 | ||
| BI010826.1 | - | 2 | 312 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000566787.1 | - | 2 | 313 | Chr. 16 - 10514842 - 10528202 [-] on Build GRCh38 | |

LINC01290
Genomic Map