siRNA Details
Gene Symbol: LOC102723322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134311.1 | 3 | 425 | |||
GenBank mRNA | BX094626.1 | - | 3 | 425 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000562577.1 | - | 3 | 425 | Chr. 8 - 85172077 - 85177041 [-] on Build GRCh38 |
