siRNA Details
| Gene Symbol: LOC102723322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134311.1 | 3 | 354 | |||
| GenBank mRNA | BX094626.1 | - | 3 | 354 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000562577.1 | - | 3 | 354 | Chr. 8 - 85172077 - 85177041 [-] on Build GRCh38 | |

LOC102723322
Genomic Map