siRNA Details
Gene Symbol: LOC105371335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135171.1 | 3 | 279 | |||
NR_135172.1 | 2 | 157 | ||||
GenBank mRNA | HY013069.1 | - | 2 | 158 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000569271.1 | - | 3 | 227 | Chr. 16 - 71513863 - 71517849 [+] on Build GRCh38 |
