siRNA Details
| Gene Symbol: LOC101927809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134605.1 | 2 | 242 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000568654.1 | - | 2 | 255 | Chr. 18 - 36179996 - 36187448 [-] on Build GRCh38 | |

LOC101927809
Genomic Map