siRNA Details
| Gene Symbol: BGLT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121648.1 | 1 | 700 | |||
| GenBank mRNA | AY034471.1 | - | 1 | 699 | ||
| KF110790.1 | - | 1 | 3377 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | U01317.3 | - | 2 | 44407 | Chr. 11 - 5215910 - 5289430 [-] on Build GRCh38 | |

BGLT3
Genomic Map