siRNA Details
| Gene Symbol: BGLT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121648.1 | 1 | 301 | |||
| GenBank mRNA | AY034471.1 | - | 1 | 301 | ||
| BU658638.1 | - | 1 | 311 | |||
| KF110790.1 | - | 1 | 2978 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BGLT3| Gene Symbol: BGLT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121648.1 | 1 | 301 | |||
| GenBank mRNA | AY034471.1 | - | 1 | 301 | ||
| BU658638.1 | - | 1 | 311 | |||
| KF110790.1 | - | 1 | 2978 | |||