siRNA Details
| Gene Symbol: LOC100653233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120505.1 | 3 | 1295 | |||
| GenBank mRNA | BC038782.1 | - | 3 | 1291 | ||
| BC048984.1 | - | 3 | 1279 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100653233| Gene Symbol: LOC100653233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120505.1 | 3 | 1295 | |||
| GenBank mRNA | BC038782.1 | - | 3 | 1291 | ||
| BC048984.1 | - | 3 | 1279 | |||