siRNA Details
| Gene Symbol: MDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092821.1 | - | 4 | 580 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000528804.1 | - | 4 | 580 | Chr. 14 - 24212739 - 24215987 [-] on Build GRCh38 | |

MDP1
Genomic Map