siRNA Details
| Gene Symbol: LINC01541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038325.1 | 7 | 2117 | |||
| NR_038326.1 | 5 | 1732 | ||||
| GenBank mRNA | AK093976.1 | - | 5 | 1717 | ||
| BX648957.1 | - | 7 | 2118 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01541| Gene Symbol: LINC01541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038325.1 | 7 | 2117 | |||
| NR_038326.1 | 5 | 1732 | ||||
| GenBank mRNA | AK093976.1 | - | 5 | 1717 | ||
| BX648957.1 | - | 7 | 2118 | |||