siRNA Details
| Gene Symbol: LINC01541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038325.1 | 7 | 2299 | |||
| NR_038326.1 | 5 | 1914 | ||||
| GenBank mRNA | BX648957.1 | - | 7 | 2300 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01541| Gene Symbol: LINC01541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038325.1 | 7 | 2299 | |||
| NR_038326.1 | 5 | 1914 | ||||
| GenBank mRNA | BX648957.1 | - | 7 | 2300 | ||