siRNA Details
| Gene Symbol: LINC01227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098656.1 | - | 1 | 559 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000567317.5 | - | 1 | 559 | Chr. 16 - 80566794 - 80569687 [-] on Build GRCh38 | |

LINC01227
Genomic Map