siRNA Details
Gene Symbol: LOC101927410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110777.1 | 3 | 2257 | |||
GenBank mRNA | AK056572.1 | - | 3 | 2257 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000562202.1 | - | 3 | 2257 | Chr. 18 - 10661933 - 10666887 [+] on Build GRCh38 |
