siRNA Details
| Gene Symbol: CASC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104152.1 | 3 | 166 | |||
| GenBank mRNA | BC039327.1 | - | 3 | 164 | ||
| BI459971.1 | - | 3 | 169 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000569074.1 | - | 3 | 164 | Chr. 17 - 71097775 - 71202177 [-] on Build GRCh38 | |

CASC17
Genomic Map