siRNA Details
| Gene Symbol: LOC101929634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135293.1 | 2 | 3228 | |||
| GenBank mRNA | AK128797.1 | - | 2 | 3225 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000564585.1 | - | 2 | 3228 | Chr. 14 - 104661120 - 104665558 [-] on Build GRCh38 | |

LOC101929634
Genomic Map