siRNA Details
Gene Symbol: LOC101928417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110937.1 | 3 | 1357 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000565238.1 | - | 3 | 1357 | Chr. 16 - 82953230 - 82990298 [-] on Build GRCh38 |
