siRNA Details
| Gene Symbol: PRCAT47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131947.1 | 4 | 460 | |||
| NR_131948.1 | 3 | 346 | ||||
| GenBank mRNA | AK093002.1 | - | 3 | 346 | ||
| BG166652.1 | - | 3 | 330 | |||
| BG171753.1 | - | 297 | ||||
| CR739737.1 | - | 2 | 283 | |||
| CR748417.1 | - | 4 | 424 | |||
| DA944068.1 | - | 3 | 346 | |||
| DA950319.1 | - | 3 | 343 | |||

PRCAT47
Genomic Map