siRNA Details
Gene Symbol: LINC01254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110775.1 | 1 | 865 | |||
GenBank mRNA | AY726567.1 | - | 1 | 865 | ||
BC040323.1 | - | 1 | 1010 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000567609.1 | - | 1 | 865 | Chr. 18 - 10405133 - 10414370 [-] on Build GRCh38 |
