siRNA Details
| Gene Symbol: LOC105370829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135679.1 | 1 | 98 | |||
| GenBank mRNA | BX103808.1 | - | 1 | 98 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000569661.5 | - | 1 | 98 | Chr. 15 - 55056747 - 55092173 [+] on Build GRCh38 | |

LOC105370829
Genomic Map