siRNA Details
| Gene Symbol: LOC105371046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135176.1 | 3 | 423 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000563162.1 | - | 3 | 423 | Chr. 16 - 1580527 - 1610328 [+] on Build GRCh38 | |

LOC105371046
Genomic Map